Skip to Content
Merck
All Photos(1)

Documents

A4673

Supelco

Albendazole

analytical standard, ≥98%

Synonym(s):

Methyl 5-(propylthio)-2-benzimidazolecarbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15N3O2S
CAS Number:
Molecular Weight:
265.33
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98%

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

CCCSc1ccc2[nH]c(NC(=O)OC)nc2c1

InChI

1S/C12H15N3O2S/c1-3-6-18-8-4-5-9-10(7-8)14-11(13-9)15-12(16)17-2/h4-5,7H,3,6H2,1-2H3,(H2,13,14,15,16)

InChI key

HXHWSAZORRCQMX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Albendazole is a benzimidazole derivative used for the treatment of human gastrointestinal helmintiasis. It inhibits microtubule and blocks the glucose uptake that causes depletion of glycogen stores and lowering formation of adinosine triphosphate (ATP) in the larval and adult stages of susceptible parasite.
Chemical structure: benzimidazole

Application

Albendazole may be used as a reference standard for the determination of albendazole and its major metabolites, albendazole sulphoxide and albendazole sulphone by high-performance liquid chromatography (HPLC) method in mice plasma.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Binds to tubulin and inhibits microtubule assembly.

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2 - STOT RE 2 Oral

Target Organs

Adrenal gland,Blood,male reproductive organs,spleen

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H D Solana et al.
Acta physiologica, pharmacologica et therapeutica latinoamericana : organo de la Asociacion Latinoamericana de Ciencias Fisiologicas y [de] la Asociacion Latinoamericana de Farmacologia, 48(4), 199-205 (1999-01-23)
Albendazole (ABZ) is an anthelmintic benzimidazole drug widely used in human and veterinary medicine. ABZ has binding affinity to both mammalian and helminth parasite tubulin. In the current work, we have performed in vitro assays and in vivo experiments in
Quantitative determination of albendazole and its main metabolites in plasma.
Garcia J J, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 723(1-2), 265-271 (1999)
Martin Spicher et al.
Antimicrobial agents and chemotherapy, 52(9), 3447-3450 (2008-07-16)
In vitro treatment of Echinococcus multilocularis and Echinococcus granulosus larval stages with the antimalarials dihydroartemisinin and artesunate (10 to 40 microM) exhibited promising results, while 6 weeks of in vivo treatment of mice infected with E. multilocularis metacestodes (200 mg/kg
Role of excipients in the crystallization of Albendazole.
Raval M K, et al.
Advanced Powder Technology, 26(4), 1102-1115 (2015)
Albendazole as a promising molecule for tumor control.
Castro L S E P W, et al.
Redox Biology, 10, 90-99 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service