Skip to Content
Merck
All Photos(1)

Key Documents

1224551

USP

Docetaxel

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Docetaxel trihydrate

Sign Into View Organizational & Contract Pricing

Select a Size

200 MG
$5,140.00

$5,140.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
200 MG
$5,140.00

About This Item

Empirical Formula (Hill Notation):
C43H53NO14 · 3H2O
CAS Number:
Molecular Weight:
861.93
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$5,140.00


Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

docetaxel

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

N([C@H]([C@@H](O)C(=O)O[C@H]2C[C@]3([C@H]([C@@H]5[C@]6([C@H](OC6)C[C@@H]([C@]5(C(=O)[C@@H](C(=C2C)C3(C)C)O)C)O)OC(=O)C)OC(=O)c4ccccc4)O)c1ccccc1)C(=O)OC(C)(C)C.O.O.O

InChI

1S/C43H53NO14.3H2O/c1-22-26(55-37(51)32(48)30(24-15-11-9-12-16-24)44-38(52)58-39(3,4)5)20-43(53)35(56-36(50)25-17-13-10-14-18-25)33-41(8,34(49)31(47)29(22)40(43,6)7)27(46)19-28-42(33,21-54-28)57-23(2)45;;;/h9-18,26-28,30-33,35,46-48,53H,19-21H2,1-8H3,(H,44,52);3*1H2/t26-,27-,28+,30-,31+,32+,33-,35-,41+,42-,43+;;;/m0.../s1

InChI key

XCDIRYDKECHIPE-QHEQPUDQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Docetaxel USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Also, for use with USP monograph such as Docetaxel Injection

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alice Dragomir et al.
BMC health services research, 14, 252-252 (2014-06-15)
For Canadian men, prostate cancer (PCa) is the most common cancer and the 3rd leading cause of cancer mortality. Men dying of PCa do so after failing castration. The management of metastatic castration-resistant prostate cancer (mCRPC) is complex and the
Fred Saad et al.
European urology, 67(2), 223-230 (2014-08-31)
Enzalutamide significantly prolonged the survival of men with metastatic castration-resistant prostate cancer (PCa) after docetaxel in the randomised, phase 3, double-blind, placebo-controlled, multinational Patients with Progressive Castration-Resistant Prostate Cancer Previously Treated with Docetaxel-Based Chemotherapy (AFFIRM) trial (NCT00974311). Prostate-specific antigen (PSA)
Josien C Haan et al.
Nature communications, 5, 5457-5457 (2014-11-15)
Response to drug therapy in individual colorectal cancer (CRC) patients is associated with tumour biology. Here we describe the genomic landscape of tumour samples of a homogeneous well-annotated series of patients with metastatic CRC (mCRC) of two phase III clinical
Monisha Sudarshan et al.
Annals of surgical oncology, 22(1), 324-330 (2014-07-16)
We have previously identified Docetaxel, Cisplatin, and 5FU (DCF) as a safe, tolerable, and effective regimen in the neoadjuvant setting for locally advanced adenocarcinoma (ADC) of the esophagus and esophagogastric junction (EGJ). We hypothesized that DCF combined with enhanced surgical
Isamu Okamoto et al.
Journal of thoracic oncology : official publication of the International Association for the Study of Lung Cancer, 10(2), 346-352 (2014-10-10)
This phase I, open-label study evaluated the safety/tolerability and maximum tolerated dose of second-line nintedanib combined with docetaxel in Japanese patients with advanced non-small-cell lung cancer. Eligible patients received docetaxel 60 or 75 mg/m(2) (day 1) plus nintedanib 100, 150

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service