Skip to Content
Merck
All Photos(1)

Key Documents

1134357

USP

Cisplatin

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

cis-Diammineplatinum(II) dichloride, cis-Dichlorodiammine platinum(II), cis-Platinum(II) diammine dichloride, Cisplatin

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$635.00

$635.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$635.00

About This Item

Linear Formula:
Pt(NH3)2Cl2
CAS Number:
Molecular Weight:
300.05
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$635.00


Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

cisplatin

manufacturer/tradename

USP

mp

270 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

N.N.Cl[Pt]Cl

InChI

1S/2ClH.2H3N.Pt/h2*1H;2*1H3;/q;;;;+2/p-2

InChI key

LXZZYRPGZAFOLE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Cisplatin USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Cisplatin Injection
  • Cisplatin for Injection
  • Carboplatin Injection

Biochem/physiol Actions

Potent platinum-based antineoplastic agent. Forms cytotoxic adducts with the DNA dinucleotide d(pGpG), inducing intrastrand cross-links.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Oral - Carc. 1B - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cisplatin
United States Pharmacopeia, 41(6), 1030-1030 (2020)
Carboplatin Injection
United States Pharmacopeia, 46(5) (2023)
Cisplatin for Injection
United States Pharmacopeia, 46(4) (2021)

Questions

  1. What is the solubility and stability after reconstitution for product 1134357, USP, Cisplatin?

    1 answer
    1. The cisplatin is soluble in water (0.25 g/100ml at 25°C), soluble in ethanol (<1 mg/ml at 25°C), and soluble in DMF

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service