Skip to Content
Merck
All Photos(1)

Key Documents

1086222

USP

Calcitonin Salmon Related Compound B

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Calcitonin salmon-glycine

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$3,760.00

$3,760.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$3,760.00

About This Item

Empirical Formula (Hill Notation):
C147H242N44O50S2
CAS Number:
Molecular Weight:
3489.89
UNSPSC Code:
41116107
NACRES:
NA.24

$3,760.00


Estimated to ship on31 May 2025


Request a Bulk Order

grade

pharmaceutical primary standard

Agency

USP

API family

calcitonin

manufacturer/tradename

USP

application(s)

pharmaceutical

format

neat

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Calcitonin Salmon is a polypeptide that has the same sequence as that of the hormone that regulates calcium metabolism and is secreted by the ultimobranchial gland of salmon. It is produced from either synthetic processes or microbial processes using recombinant DNA (rDNA) technology. The host cell-derived protein content and the host cell- or vector-derived DNA content of Calcitonin Salmon produced from an rDNA process are determined by validated methods. This product contains NLT 90.0% and NMT 105.0% of calcitonin salmon, calculated on an acetic acid-free and anhydrous basis.[1]

Calcitonin Salmon USP reference standard is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Calcitonin Salmon Related Compound B USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monograph such as Calcitonin Salmon

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

This product is part of USP Biologics Program.
Sales restrictions may apply.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Calcitonin Salmon
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 36(5), 670-670 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service