Skip to Content
Merck
All Photos(1)

Key Documents

Y0000696

Testosterone decanoate for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Testosterone decanoate, 17β-Hydroxy-4-androsten-3-one 17-decanoate, 4-Androsten-17β-ol-3-one 17-caprate

Sign Into View Organizational & Contract Pricing

Select a Size

0.4 MG
$204.00

$204.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
0.4 MG
$204.00

About This Item

Empirical Formula (Hill Notation):
C29H46O3
CAS Number:
Molecular Weight:
442.67
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$204.00


Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

testosterone

manufacturer/tradename

EDQM

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

CCCCCCCCCC(=O)O[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C

InChI

1S/C29H46O3/c1-4-5-6-7-8-9-10-11-27(31)32-26-15-14-24-23-13-12-21-20-22(30)16-18-28(21,2)25(23)17-19-29(24,26)3/h20,23-26H,4-19H2,1-3H3/t23-,24-,25-,26-,28-,29-/m0/s1

InChI key

LBERVHLCXUMDOT-MPZZESAYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Testosterone decanoate for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Carc. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patricia Regal et al.
Journal of agricultural and food chemistry, 63(1), 370-378 (2014-12-09)
The endogenous occurrence of natural hormones obstructs the application of classical targeted methods as confirmatory options. In the case of estradiol, the ultimate confirmation of its exogenous administration relies on gas chromatography coupled to combustion/isotope ratio mass spectrometry (GC-C/IRMS). A

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service