Y0002306
Nebivolol for peak identification
European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Nebivolol
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
nebivolol
manufacturer/tradename
EDQM
storage temp.
2-8°C
SMILES string
Fc1cc2c(cc1)O[C@H](CC2)[C@@H](O)CNC[C@H](O)[C@H]3Oc4c(cc(cc4)F)CC3
InChI
1S/C22H25F2NO4/c23-15-3-7-19-13(9-15)1-5-21(28-19)17(26)11-25-12-18(27)22-6-2-14-10-16(24)4-8-20(14)29-22/h3-4,7-10,17-18,21-22,25-27H,1-2,5-6,11-12H2/t17-,18-,21-,22+/m0/s1
InChI key
KOHIRBRYDXPAMZ-YHDSQAASSA-N
General description
Application
Packaging
Other Notes
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service