Skip to Content
Merck
All Photos(1)

Key Documents

PHR1623

Supelco

Galantamine Hydrobromide

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Galantamine hydrobromide, Galanthamine hydrobromide

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$503.00

$503.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$503.00

About This Item

Empirical Formula (Hill Notation):
C17H21NO3 · HBr
CAS Number:
Molecular Weight:
368.27
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$503.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. Y0001279
traceable to USP 1287755

API family

galantamine

CofA

current certificate can be downloaded

packaging

pkg of 500 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

[H][C@]1(O2)C[C@@H](O)C=C[C@@]13C4=C2C(OC)=CC=C4CN(C)CC3.Br

InChI

1S/C17H21NO3.BrH/c1-18-8-7-17-6-5-12(19)9-14(17)21-16-13(20-2)4-3-11(10-18)15(16)17;/h3-6,12,14,19H,7-10H2,1-2H3;1H/t12-,14-,17-;/m0./s1

InChI key

QORVDGQLPPAFRS-XPSHAMGMSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Galantamine Hydrobromide, a reversible, competitive acetyl cholinesterase inhibitor, is suggested for the remedy of mild to moderate dementia of the Alzheimer′s type.[1][2]

Application

Galantamine Hydrobromide may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by various chromatography techniques.[1][2][3][4]
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC1681 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantitative determination of galantamine hydrobromide in pharmaceutical dosage form by RP-high performance liquid chromatography.
Krupesh BP, et al.
Journal of Chemical and Pharmaceutical Research, 2(2), 36-43 (2010)
Improving method capability of a drug substance HPLC assay.
Dejaegher B, et al.
Journal of Pharmaceutical and Biomedical Analysis, 42(2), 155-170 (2006)
Pharmacokinetics and bioequivalence studies of galantamine hydrobromide dispersible tablet in healthy male Chinese volunteers.
Zhang LJ, et al.
Drug Development and Industrial Pharmacy, 33(3) (2007)
Oil-in-water microemulsion high performance liquid chromatographic analysis of pharmaceuticals.
Marsh A, et al.
Chromatographia, 59(9), 531-542 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service