Skip to Content
Merck
All Photos(1)

Key Documents

Y0002224

Sorafenib tosilate

CRS, European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Sorafenib tosylate, 4-[4-[[[[4-Chloro-3-(trifluoromethyl)phenyl]amino]carbonyl]amino]phenoxy]-N-methyl-2-pyridinecarboxamide, 4-methylbenzenesulfonate, BAY 43-9006 tosylate salt, BAY43-9006 tosylate salt, N-[4-Chloro-3-(trifluoromethyl)phenyl]-N′-[4-[2-(N-methylcarbamoyl)-4-pyridyloxy]phenyl]urea, 4-methylbenzenesulfonate

Sign Into View Organizational & Contract Pricing

Select a Size

120 MG
$292.00

$292.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
120 MG
$292.00

About This Item

Empirical Formula (Hill Notation):
C21H16ClF3N4O3·C7H8SO3
CAS Number:
Molecular Weight:
637.03
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$292.00


Check Cart for Availability

Request a Bulk Order

API family

sorafenib

manufacturer/tradename

EDQM

storage temp.

2-8°C

SMILES string

FC(F)(F)c2c(ccc(c2)NC(=O)Nc3ccc(cc3)Oc4cc([n+H]cc4)C(=O)NC)Cl.[S](=O)(=O)([O-])c1ccc(cc1)C

InChI

1S/C21H16ClF3N4O3.C7H8O3S/c1-26-19(30)18-11-15(8-9-27-18)32-14-5-2-12(3-6-14)28-20(31)29-13-4-7-17(22)16(10-13)21(23,24)25;1-6-2-4-7(5-3-6)11(8,9)10/h2-11H,1H3,(H,26,30)(H2,28,29,31);2-5H,1H3,(H,8,9,10)

InChI key

IVDHYUQIDRJSTI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Sorafenib tosilate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Orally active kinase inhibitor that exerts anticancer efficacy via targeting Raf-1 (c-Raf), b-Raf, VEGFR-2 (Flk-1, KDR), VEGFR-3, PDGFR-β, Flt-3, c-KIT, FGFR-1 (Flt-2) and RET.
Tosylate salt of the orally active kinase inhibitor Sorafenib (BAY 43-9006) that exerts broad-spectrum anticancer efficacy in vitro and in vivo via targeting b-Raf, c-Raf (Raf-1), as well as several receptor tyrosine kinases involved in neovascularization and tumor progression, including vascular endothelial growth factor receptors 2/3 (VEGFR-2/Flk-1/KDR, VEGFR-3), platelet-derived growth factor receptor-beta (PDGFR-β), Flt-3, c-KIT, FGFR-1 (Flt-2) and RET.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Lact. - Repr. 1B - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service