Skip to Content
Merck
All Photos(1)

Key Documents

SRP0702

Sigma-Aldrich

SORAFENIB

recombinant

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$129.00

$129.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$129.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

$129.00


Check Cart for Availability

Request a Bulk Order

Assay

>95% (TLC)

form

lyophilized powder

mol wt

~464.8

shipped in

ambient

storage temp.

−20°C

Application

Clinically useful anticancer agent.

Biochem/physiol Actions

Initially developed as a Raf kinase inhibitor, IC50 = 6 nM, but has been shown to inhibit many receptor tyrosine kinases including BRAF (IC50 = 22 nM); VEGFR-2 (IC50 = 90 nM); VEGFR-3 (IC50 = 20 nM); PDGFR-beta (IC50 = 57 nM); Flt3 (IC50 = 58 nM); c-KIT (IC50 = 68 nM); FGFR-1 (IC50 = 580 nM) (1). Paradoxically more potent in a cellular assay (IC50 = 20 nM) compared to an isolated enzyme assay (IC50 = 107 nM) for c-Fms (2). Inhibits activation of MAPK pathway and ERK phosphorylation (3). Induces caspase-independent apoptosis in melanoma cells (4).

Reconstitution

Formulated as a lyophilized powder. May be dissolved in DMSO (200 mg/mL) or Ethanol (3 mg/mL).

Storage and Stability

Store desiccated as supplied at -20°C for up to 2 years. Store solutions at -20°C for up to 1 month.

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Lact. - Repr. 1B - STOT RE 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bei Zhou et al.
Molecular therapy. Nucleic acids, 26, 1051-1065 (2021-11-18)
Emerging evidence has shown that endoplasmic reticulum (ER) stress promotes sorafenib resistance in hepatocellular carcinoma (HCC). However, the underlying mechanisms are poorly understood. The purpose of this study was to explore the mechanism by which ER stress promotes sorafenib resistance

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service