Skip to Content
Merck
All Photos(1)

Key Documents

SAAR1591100

Charcoal

powder (activated), uniTEK

Synonym(s):

Activated charcoal, Charcoal activated

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
MDL number:
UNSPSC Code:
11101522

vapor pressure

<0.1 mmHg ( 20 °C)

form

powder (activated)

autoignition temp.

842 °F

packaging

poly bottle of 1 × 500 g (SAAR1591100EM)

resistivity

1375 μΩ-cm, 20°C (graphite)

mp

3550 °C (lit.)

storage temp.

2-30°C

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

For laboratory use only

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service