Skip to Content
Merck
All Photos(1)

Key Documents

1.02186

Supelco

Charcoal activated

for analysis

Synonym(s):

Charcoal activated

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
$287.00
20 KG
$2,190.00

$287.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
1 KG
$287.00
20 KG
$2,190.00

About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
MDL number:
UNSPSC Code:
11101522
EC Index Number:
231-153-3
NACRES:
NB.21

$287.00


Estimated to ship on29 May 2025


Request a Bulk Order

Quality Level

form

solid

surface area

800-1500 m2/g

impurities

≤0.2% EtOH solubles
≤0.5% Substances soluble in water
≤0.5% water solubles
≤1% HNO3 solubles

ign. residue

≤1% (600 °C)

loss

≤ 10% loss on drying

particle size

<100 μm (~90%)

mp

3550 °C

bulk density

150‑440 kg/m3

anion traces

sulfate (SO42-): ≤100 ppm

cation traces

Fe: ≤300 ppm
Pb: ≤10 ppm
Zn: ≤10 ppm

storage temp.

no temp limit

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Analysis Note

Identity: conforms
Substances soluble in nitric acid: ≤ 1 %
Substances soluble in water: ≤ 0.5 %
Substances soluble in Ethanol: ≤ 0.2 %
Sulphate (SO₄): ≤ 100 ppm
Fe (Iron): ≤ 300 ppm
Pb (Lead): ≤ 10 ppm
Zn (Zinc): ≤ 10 ppm
n-Hexane adsorption: ≥ 30 %
Residue on ignition (600 °C): ≤ 1 %
Loss on drying: ≤ 10 %
Particle size (< 100 µm): about 90 %

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service