Skip to Content
Merck
All Photos(1)

Key Documents

SAAR1461090

Buffer solution

pH 9.00±0.02 (20 °C±0.1)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

packaging

poly bottle of 1 × 500 mL (SAAR1461090KF)

pH

9.00±0.02 (20 °C±0.1)

storage temp.

15-25°C

Application

For laboratory use only

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service