Skip to Content
Merck
All Photos(1)

Key Documents

PHL86291

Albine hydrochloride

phyproof® Reference Substance

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$2,270.00

$2,270.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$2,270.00

About This Item

Empirical Formula (Hill Notation):
C14H20N2O · HCl
CAS Number:
Molecular Weight:
268.78
UNSPSC Code:
41116107
NACRES:
NA.24

$2,270.00


Check Cart for Availability

Request a Bulk Order

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥90.0% (HPLC)

manufacturer/tradename

PhytoLab

format

neat

storage temp.

−20°C

SMILES string

O=C1C=CN2[C@]([C@]3([H])[C@@H](CC=C)NC[C@@]([H])(C3)C2)([H])C1.Cl

InChI

1S/C14H20N2O/c1-2-3-13-12-6-10(8-15-13)9-16-5-4-11(17)7-14(12)16/h2,4-5,10,12-15H,1,3,6-9H2/t10-,12+,13+,14+/m0/s1

InChI key

QJVOZXGJOGJKPT-IGHBBLSQSA-N

Related Categories

General description

This substance is supplied with certified chromatographic purity. The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service