Skip to Content
Merck
All Photos(1)

Key Documents

92052

Supelco

(+)-Sparteine

≥98.0% (GC)

Synonym(s):

Pachycarpine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$267.00

$267.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
1 G
$267.00

About This Item

Empirical Formula (Hill Notation):
C15H26N2
CAS Number:
Molecular Weight:
234.38
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$267.00


Estimated to ship on01 June 2025


Request a Bulk Order

Quality Level

Assay

≥98.0% (GC)

form

liquid

optical activity

[α]/D +17.0±1.5°, c = 1 in ethanol

SMILES string

[H][C@@]1([C@@]2([H])CN3CCCC[C@@]3([H])[C@@](C2)([H])C4)N4CCCC1

InChI

1S/C15H26N2/c1-3-7-16-11-13-9-12(14(16)5-1)10-17-8-4-2-6-15(13)17/h12-15H,1-11H2/t12-,13-,14-,15+/m1/s1

InChI key

SLRCCWJSBJZJBV-TUVASFSCSA-N

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of enantiomerically pure 2,2'-dihydroxy-1,1'-binaphthyl, 2,2'-diamino-1,1'-binaphthyl, and 2-amino-2'-hydroxy-1,1'-binaphthyl. Comparison of processes operating as diastereoselective crystallization and as second order asymmetric transformation.
Smrcina, M,. et al.
The Journal of Organic Chemistry, 57, 1917-1920 (1992)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service