Skip to Content
Merck
All Photos(1)

Key Documents

PHL84245

Notoginsenoside R2

phyproof® Reference Substance

Synonym(s):

Ginsenoside Ng-R2

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$703.00

$703.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$703.00

About This Item

Empirical Formula (Hill Notation):
C41H70O13
CAS Number:
Molecular Weight:
770.99
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$703.00


Check Cart for Availability

Request a Bulk Order

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥90.0% (HPLC)

form

solid

manufacturer/tradename

PhytoLab

SMILES string

O1[C@H]([C@@H]([C@H]([C@@H]([C@H]1CO)O)O)O[C@@H]6OC[C@H]([C@@H]([C@H]6O)O)O)O[C@@H]2[C@@H]3[C@@]([C@@H]4[C@]([C@]5([C@H]([C@@H](C4)O)[C@H](CC5)[C@@](O)(CCC=C(C)C)C)C)(C2)C)(CC[C@@H](C3(C)C)O)C

InChI

1S/C41H70O13/c1-20(2)10-9-13-41(8,50)21-11-15-39(6)28(21)22(43)16-26-38(5)14-12-27(45)37(3,4)34(38)24(17-40(26,39)7)52-36-33(31(48)30(47)25(18-42)53-36)54-35-32(49)29(46)23(44)19-51-35/h10,21-36,42-50H,9,11-19H2,1-8H3/t21-,22+,23+,24-,25+,26+,27-,28-,29-,

InChI key

FNIRVWPHRMMRQI-PGOMJGFXSA-N

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Other Notes

This compound is commonly found in plants of the genus: panax

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service