Skip to Content
Merck
All Photos(1)

Key Documents

CRM78340

Supelco

1-Octene

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$196.00

$196.00


Check Cart for Availability


Select a Size

Change View
100 MG
$196.00

About This Item

Linear Formula:
CH3(CH2)5CH=CH2
CAS Number:
Molecular Weight:
112.21
Beilstein:
1734497
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$196.00


Check Cart for Availability

grade

certified reference material
TraceCERT®

Quality Level

vapor density

3.9 (vs air)

vapor pressure

36 mmHg ( 38 °C)

product line

TraceCERT®

autoignition temp.

446 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

3.9 %

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

refractive index

n20/D 1.408 (lit.)
n20/D 1.409 (lit.)

bp

122-123 °C (lit.)

mp

−101 °C (lit.)

density

0.715 g/mL at 25 °C (lit.)

application(s)

petroleum

storage temp.

−20°C

SMILES string

CCCCCCC=C

InChI

1S/C8H16/c1-3-5-7-8-6-4-2/h3H,1,4-8H2,2H3

InChI key

KWKAKUADMBZCLK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

50.0 °F - closed cup

Flash Point(C)

10 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service