Skip to Content
Merck
All Photos(1)

Key Documents

8.20928

Sigma-Aldrich

1-Octene

for synthesis

Synonym(s):

1-Octene, 1-Octylene, Caprylene

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$39.20
250 ML
$47.70
1 L
$89.10

$39.20


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
5 ML
$39.20
250 ML
$47.70
1 L
$89.10

About This Item

Linear Formula:
CH3(CH2)5CH=CH2
CAS Number:
Molecular Weight:
112.21
MDL number:
UNSPSC Code:
12352002
EC Index Number:
203-893-7
NACRES:
NA.22

$39.20


Estimated to ship on29 May 2025


Request a Bulk Order

vapor pressure

17.3 hPa ( 20 °C)

Quality Level

Assay

≥97.0% (GC)

form

liquid

autoignition temp.

221 °C

potency

>5000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

expl. lim.

0.7-6.8 % (v/v)

kinematic viscosity

0.38 cSt(40 °C)

mp

-102 °C

transition temp

flash point 8 °C

density

0.72 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

C(CCC)CCC=C

InChI

1S/C8H16/c1-3-5-7-8-6-4-2/h3H,1,4-8H2,2H3

InChI key

KWKAKUADMBZCLK-UHFFFAOYSA-N

Application

  • Catalytic Behavior of Chromium: Investigation of chromium catalysts for selective ethylene oligomerization to 1-Octene, crucial for developing advanced materials and chemicals used in various sectors, including pharmaceuticals (KA Alferov et al., 2017).
  • Analysis of Ethylene/1-Octene Copolymers: Detailed structure analysis of ethylene/1-Octene copolymers synthesized from living coordination polymerization, providing insights into the polymer structure that are critical for developing new materials (W Liu et al., 2014).
  • Phenoxyimine Zr-Catalyzed Polymerizations: Studies on unexpected s-ligand effects in phenoxyimine Zr-catalyzed copolymerizations of ethylene and 1-Octene, relevant for designing catalysts and polymers with specific properties for drug delivery systems (Y Gao et al., 2019).

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.714 - 0.715
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

46.4 °F

Flash Point(C)

8 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service