Skip to Content
Merck
All Photos(1)

Key Documents

CRM33859

Supelco

Metolachlor, predominantly S-Metolachlor

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

2-Chloro-N-(2-ethyl-6-methylphenyl)-N-[(1S)-2-methoxy-1-methylethyl]acetamide

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$130.00

$130.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$130.00

About This Item

Empirical Formula (Hill Notation):
C15H22ClNO2
CAS Number:
Molecular Weight:
283.79
Beilstein:
6483536
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$130.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

format

neat

storage temp.

−20°C

SMILES string

CCc1cccc(C)c1N([C@@H](C)COC)C(=O)CCl

InChI

1S/C15H22ClNO2/c1-5-13-8-6-7-11(2)15(13)17(14(18)9-16)12(3)10-19-4/h6-8,12H,5,9-10H2,1-4H3/t12-/m0/s1

InChI key

WVQBLGZPHOPPFO-LBPRGKRZSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.Download your certificate at: http://www.sigma-aldrich.com

Application

Environmental
S-Metolachlor is a selective herbicide, absorbed primarily by hypocotyls and shoots. It is used as pre-, early post-emergence and pre-plant herbicide, incorporated to control annual grass and some broad-leaved weeds in maize, sorghum, cotton, surgar beets. Under Reg. (EC) No 1107/2009 S-Metolachlor is approved for use as a plant protection product in the European Union (EU). Maximum residue levels (MRLs) have been set according Reg. (EU) No 1317/2013 for metolachlor and S-metolachlor (metolachlor including other mixtures of constituent isomers including S-metolachlor) for various products of plant and animal origin from 0.01 - 0.05 mg/kg. Metolachlor (technical product being an isomeric mixture of R- and S-enantiomers) is not approved for use as a plant protection product in the European Union (EU) or UK. The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service