Skip to Content
Merck
All Photos(1)

Key Documents

32059

Supelco

Simazine

PESTANAL®, analytical standard

Synonym(s):

2,4-Bis(ethylamino)-6-chloro-1,3,5-triazine

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$141.00

$141.00


Available to ship on07 April 2025Details


Request a Bulk Order

Select a Size

Change View
250 MG
$141.00

About This Item

Empirical Formula (Hill Notation):
C7H12ClN5
CAS Number:
Molecular Weight:
201.66
Beilstein:
10895
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$141.00


Available to ship on07 April 2025Details


Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

CCNc1nc(Cl)nc(NCC)n1

InChI

1S/C7H12ClN5/c1-3-9-6-11-5(8)12-7(13-6)10-4-2/h3-4H2,1-2H3,(H2,9,10,11,12,13)

InChI key

ODCWYMIRDDJXKW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Simazine is a triazine herbicide mostly used in corn production.[1] It can be converted by corn seedling to hydroxysimazine, which is a detoxified form.[2]
Standard for Supelco MIP SPE cartridges. For more information request Supelco Literature T407075, T706023

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Simazine has been used as working standard in HPLC/DAD and GC/FID methods to identify and quantify triazine herbicides from water samples.[3]

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Volatilization and wind erosion of soil surface applied atrazine, simazine, alachlor, and toxaphene.
Glotfelty, Dwight E., et al.
Journal of Agricultural and Food Chemistry, 37.2, 546-551 (1989)
Simazine: Degradation by Corn Seedlings.
Hamilton RH, Moreland DE.
Science, 135(3501), 373-374 (1962)
Development and optimization of a method for the determination of simazine, atrazine and propazine using solid-phase extraction and HPLC/GC.
Trajkovska, Vera, S. Petrovska-Jovanovic, and M. I. R. K. O. Cvetkovski.
J. Serb. Chem. Soc., 66.3, 199-204 (2001)
Alzbeta Stara et al.
Environmental toxicology and pharmacology, 33(2), 334-343 (2012-02-04)
We investigated the chronic effect of simazine, an s-triazine herbicide commonly present in aquatic environments, on the antioxidant system and oxidative stress indices in common carp (Cyprinus carpio L.). Fish were exposed to sub-lethal concentrations of 0.06 μg l(-1) (environmental
Guixiang Zhang et al.
Environmental pollution (Barking, Essex : 1987), 159(10), 2594-2601 (2011-07-02)
Simazine sorption to corn straw biochars prepared at various temperatures (100-600 °C) was examined to understand its sorption behavior as influenced by characteristics of biochars. Biochars were characterized via elemental analysis, BET-N(2) surface area (SA), FTIR and (13)C NMR. Freundlich

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service