Skip to Content
Merck
All Photos(1)

Key Documents

95091

Supelco

Pinoxaden Metabolite SYN552804

Synonym(s):

[4-(7,9-Dioxohexahydro-7H-pyrazolo[1,2- d][l,4,5]oxadiazepin-8-yl)-3,5-diethylphenyl]methyl D-glucopyranoside

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$128.00

$128.00


Estimated to ship on17 April 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$128.00

About This Item

Empirical Formula (Hill Notation):
C24H34N2O9
Molecular Weight:
494.53
UNSPSC Code:
41116107
NACRES:
NA.24

$128.00


Estimated to ship on17 April 2025


Request a Bulk Order

product line

PESTANAL®

Quality Level

shelf life

limited shelf life, expiry date on the label

format

neat

storage temp.

2-8°C

Related Categories

Application

Environmental

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 2 - Skin Sens. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service