Skip to Content
Merck
All Photos(1)

Key Documents

89968

Supelco

2,2′-Methylenebis(6-tert-butyl-4-methylphenol)

analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$163.00

$163.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$163.00

About This Item

Linear Formula:
CH2[C6H2[C(CH3)3](CH3)OH]2
CAS Number:
Molecular Weight:
340.50
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$163.00


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥98.5% (GC)

form

crystals

shelf life

limited shelf life, expiry date on the label

mp

123-127 °C (lit.)

SMILES string

Cc1cc(Cc2cc(C)cc(c2O)C(C)(C)C)c(O)c(c1)C(C)(C)C

InChI

1S/C23H32O2/c1-14-9-16(20(24)18(11-14)22(3,4)5)13-17-10-15(2)12-19(21(17)25)23(6,7)8/h9-12,24-25H,13H2,1-8H3

InChI key

KGRVJHAUYBGFFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Find all available reference materials for compounds listed in 10/2011 here

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

365.0 °F - Pensky-Martens closed cup

Flash Point(C)

185 °C - Pensky-Martens closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service