Skip to Content
Merck
All Photos(1)

Key Documents

72818

Supelco

Methylhydroquinone

analytical standard

Synonym(s):

Toluhydroquinone

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$96.40

$96.40


Estimated to ship on08 April 2025



Select a Size

Change View
100 MG
$96.40

About This Item

Linear Formula:
CH3C6H3-1,4-(OH)2
CAS Number:
Molecular Weight:
124.14
Beilstein:
2041489
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.24

$96.40


Estimated to ship on08 April 2025


grade

analytical standard

Quality Level

Assay

≥99%

autoignition temp.

851 °F

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

128-130 °C (lit.)

format

neat

storage temp.

2-30°C

SMILES string

Cc1cc(O)ccc1O

InChI

1S/C7H8O2/c1-5-4-6(8)2-3-7(5)9/h2-4,8-9H,1H3

InChI key

CNHDIAIOKMXOLK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1A - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

341.6 °F - closed cup

Flash Point(C)

172 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service