Skip to Content
Merck
All Photos(1)

Key Documents

89869

Supelco

2,6-Diaminopyridine

analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$101.00

$101.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 MG
$101.00

About This Item

Empirical Formula (Hill Notation):
C5H7N3
CAS Number:
Molecular Weight:
109.13
Beilstein:
108513
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.21

$101.00


Estimated to ship on28 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

285 °C (lit.)

mp

117-122 °C (lit.)

format

neat

storage temp.

2-30°C

SMILES string

Nc1cccc(N)n1

InChI

1S/C5H7N3/c6-4-2-1-3-5(7)8-4/h1-3H,(H4,6,7,8)

InChI key

VHNQIURBCCNWDN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

311.0 °F - closed cup

Flash Point(C)

155 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service