![2,6-Diaminopyridine analytical standard](/deepweb/assets/sigmaaldrich/product/structures/590/016/7527301e-5178-428f-86e9-c13ffc967f68/640/7527301e-5178-428f-86e9-c13ffc967f68.png)
2,6-Diaminopyridine
![2,6-Diaminopyridine analytical standard](/deepweb/assets/sigmaaldrich/product/structures/590/016/7527301e-5178-428f-86e9-c13ffc967f68/640/7527301e-5178-428f-86e9-c13ffc967f68.png)
Synonym(s):
2,6-Diaminopyridine
Linear Formula:
2,6-(NH2)2C5H3N
CAS Number:
Molecular Weight:
109.13
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing