861339
EPA 8260 Calibration Mix 1
certified reference material, 2000 μg/mL each component in methanol
About This Item
Recommended Products
grade
certified reference material
TraceCERT®
Agency
EPA 8260
description
Separate Source
product line
TraceCERT®
CofA
current certificate can be downloaded
packaging
ampule of 1.5 mL
concentration
2000 μg/mL each component in methanol
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
environmental
format
multi-component solution
storage temp.
2-8°C
Application
Other Notes
Legal Information
Analyte
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Carc. 1A - Flam. Liq. 2 - Muta. 1B - Ozone 1 - STOT RE 2 - STOT SE 1
Target Organs
Eyes,Central nervous system, Liver,Kidney
WGK
WGK 3
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service