Skip to Content
Merck
All Photos(1)

Key Documents

806727

TSKgel® G2000SW (13 µm) HPLC Columns

L x I.D. 30 cm × 21.5 mm, diol phase

Synonym(s):

TSKgel® Size Exclusion (SW-Type) HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
eCl@ss:
32110501
L × I.D.:
30 cm × 21.5 mm
particle size:
13 μm
matrix active group:
diol phase
pore size:
125 Å
matrix:
silica particle platform
Product 806727 is not currently sold in your country. Contact Technical Service

Product Name

TSKgel® G2000SW HPLC Column, phase diol, L × I.D. 30 cm × 21.5 mm, 13 μm particle size

material

stainless steel hardware

Agency

suitable for USP L20
suitable for USP L59

product line

TSKgel®

manufacturer/tradename

Tosoh 06727

parameter

≤0.5 M salt concentration
0-100% aqueous soluble organic solvents
10 bar max. pressure
10-30 °C temperature
8 mL/min flow rate

technique(s)

HPLC: suitable

L × I.D.

30 cm × 21.5 mm

matrix

silica particle platform

matrix active group

diol phase

particle size

13 μm

pore size

125 Å

operating pH

2.5-7.5

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

General description

As the first generation of silica-based SEC columns, TSKgel G2000SW provides excellent separation of peptides and proteins with molecular weights up to 100,000 Da. TSKgel G2000SW columns start at 10μm for analytical columns and also scale to 13 and 20μm in columns up to 55mm ID for preparative purifications.

Physical form

Shipped in 0.05% NaN3 and 0.1 mol/L Na2SO4 in 0.1 M phosphate buffer, pH 6.7.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service