Skip to Content
Merck
All Photos(1)

Documents

805758

TSKgel® SW Type (13 µm) HPLC Column

L x I.D. 7.5 cm × 21.5 mm, diol phase

Synonym(s):

TSKgel® Size Exclusion (SW-Type) HPLC Guard Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501

product name

TSKgel® SW Type Guard Column, phase diol, L × I.D. 7.5 cm × 21.5 mm, 13 μm particle size

material

stainless steel

Agency

suitable for USP L20
suitable for USP L59

product line

TSKgel®

manufacturer/tradename

Tosoh 05758

parameter

≤0.5 M salt concentration
0-100% aqueous soluble organic solvents
10-30 °C temperature
8 mL/min flow rate

technique(s)

HPLC: suitable

L × I.D.

7.5 cm × 21.5 mm

matrix

silica particle platform

matrix active group

diol phase

particle size

13 μm

pore size

250 Å

operating pH

2.5-7.5

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

General description

Guard Column for 21.5mmID TSKgel G2000SW, G3000SW and G4000SW columns
TSKgel Guard Columns are stand-alone and do not require separate holders.

Physical form

Shipped in 0.05% NaN3 and 0.1 mol/L Na2SO4 in 0.1 M phosphate buffer, pH 6.7.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service