Skip to Content
Merck
All Photos(1)

Key Documents

72506

Supelco

2,6-Di-tert-butyl-4-(dimethylaminomethyl)phenol

analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$83.90

$83.90


Check Cart for Availability


Select a Size

Change View
100 MG
$83.90

About This Item

Linear Formula:
(CH3)2NCH2C6H2[C(CH3)3]2OH
CAS Number:
Molecular Weight:
263.42
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$83.90


Check Cart for Availability

grade

analytical standard

Quality Level

Assay

≥97% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

172 °C/30 mmHg (lit.)

mp

90-95 °C
91-96 °C

format

neat

storage temp.

2-30°C

SMILES string

CN(C)Cc1cc(c(O)c(c1)C(C)(C)C)C(C)(C)C

InChI

1S/C17H29NO/c1-16(2,3)13-9-12(11-18(7)8)10-14(15(13)19)17(4,5)6/h9-10,19H,11H2,1-8H3

InChI key

VMZVBRIIHDRYGK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service