2,6-Di-tert-butyl-4-(dimethylaminomethyl)phenol
Linear Formula:
(CH3)2NCH2C6H2[C(CH3)3]2OH
CAS Number:
Molecular Weight:
263.42
Products
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Markush Class
Markush Group
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing