Skip to Content
Merck
All Photos(1)

Key Documents

66896-U

Supelco

Empore SPE Disks

Carbon, diam. 47 mm, pk of 20

Synonym(s):

carbon, SPE disks, 47 mm

Sign Into View Organizational & Contract Pricing

Select a Size

20 EA
$946.00

$946.00


Available to ship on22 April 2025Details


Request a Bulk OrderRequest a Sample

Select a Size

Change View
20 EA
$946.00

About This Item

UNSPSC Code:
13111000
NACRES:
NB.21

$946.00


Available to ship on22 April 2025Details


Request a Bulk OrderRequest a Sample

Product Name

Empore SPE Disks, matrix active group Carbon, diam. 47 mm, pk of 20

product line

Empore

Quality Level

composition

bed wt., 500 mg (per 47 mm disk)

packaging

pk of 20

technique(s)

solid phase extraction (SPE): suitable

diam.

47 mm

membrane thickness

0.5 mm

size

surface area

1100 m2/g

matrix

activated carbon particle platform

matrix active group

Carbon

particle size

12 μm

Looking for similar products? Visit Product Comparison Guide

General description

Empore membrane SPE technology comprises of SPE particles tightly enmeshed within a network of inert PTFE fibrils (90% SPE particles : 10% PTFE, by weight). The SPE-membrane fabrication process results in a highly dense and uniform extraction medium that offers distinct advantages over traditional sorbent/packed-bed SPE products. Empore SPE technology provides a denser, more uniform extraction bed than traditional packed bed products allowing for smaller bed weights, shorter analyte to pore diffusion paths, and more efficient extractions.

The Empore line of SPE disks comprises of the most complete line of SPE disks for extracting large volumes of aqueous samples. The product line ranges from time-tested C18 to unique phase chemistries such as carbon and the highly polar Oil & Grease disk. The disks are ideal for environmental analysis where 1 L sample volumes are not uncommon and provide an efficient alternative to LLE.

Save Time & Money:
  • Reduced SPE Bed Mass = Reduced SPE Solvent & Elution Volumes
  • Minimize SPE eluate evaporation time
  • Potentially allows for direct injection of the SPE eluate
  • Dense & Uniform Extraction Medium = No SPE Channeling and Voiding
  • Efficient mass-transfer kinetics allow for faster flow rates
  • Eliminate SPE fines improving column and instrument life

Application

Developed for the extraction of volatile and highly polar compounds from aqueous samples. Applications include:
  • Ground water - N-nitrosdeimethylamine
  • Waste water - Oxamyl and methamidophos
  • Drinking water - Trihalomethanes
  • Reagent water - Clean-up for purge and trap applications
  • Forensics - Fire accelerants, combustible vapors

Legal Information

Empore is a trademark of CDS Analytical

Disclaimer

CDS Analytical LLC recently acquired the rights to Empore products from 3M Purification Inc. For an interim phase, you may receive products packaged with either manfacturer′s branding.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service