WHA67221001
Whatman® VACU-GUARD® inline filter devices
VACU-GUARD 150, activated carbon, 1/pk
Synonym(s):
Z249564, filter devices, filtration device, inline filtration device
Select a Size
$307.00
Available to ship on28 April 2025Details
Select a Size
About This Item
$307.00
Available to ship on28 April 2025Details
Recommended Products
material
PEEK/PTFE filter membrane (PFTE filteration media)
polypropylene housing
sterility
non-sterile
feature
Trap media (Activated carbon)
packaging
pack of 1
manufacturer/tradename
Whatman 6722-1001
Whatman Article No. 28420216 (US reference)
parameter
210-450 L/min flow rate at 1.45-7.25 psi
size
9.3 in. (236 mm) , overall
surface area
82,000 m2
Looking for similar products? Visit Product Comparison Guide
General description
Application
- Vacuum pump protection
Features and Benefits
- Excellent for protecting vacuum pump systems from solvent vapor or gaseous contaminants and aqueous aerosols.
- Designed for in-line use with stepped barb connections for 1NA to 12 mm i.d. hose.
- Available with choice of chemical trap: activated carbon, molecular sieve, or desiccant.
- Membrane retains 99.99% of airbone particles greater than NA.1 µm.
- Features hydrophobic polytetrafluoroethylene (PTFE) membrane.
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service