63289-U
BIOshell lgG 1000 Å C4 (2.7 μm) HPLC Columns
L × I.D. 15 cm × 2.1 mm, HPLC Column
Synonym(s):
High Resolution IgG HPLC Column
Select a Size
$1,820.00
Select a Size
About This Item
$1,820.00
Recommended Products
Product Name
BIOshell lgG 1000 Å C4, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 15 cm × 2.1 mm
material
stainless steel hardware
Agency
suitable for USP L26
description
Shell thickness (0.5 μm)
Solid core (1.7 μm)
product line
BIOshell
feature
endcapped
specific analyte(s)
IgG
manufacturer/tradename
BIOshell
packaging
1 ea of
extent of labeling
0.6% carbon loading
parameter
1000 bar pressure (14500 psi)
90 °C max. temp.
technique(s)
HPLC: suitable
LC/MS: suitable
UHPLC-MS: suitable
UHPLC: suitable
L × I.D.
15 cm × 2.1 mm
surface area
22 m2/g
matrix
spherical silica particle platform
superficially porous particle
matrix active group
C4 (butyl) phase
particle size
2.7 μm
pore size
1000 Å
operating pH range
2-9
separation technique
reversed phase
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Legal Information
antibody
guard cartridge
Storage Class Code
13 - Non Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
BIOshell™ IgG 1000 Å C4 columns are highly suited for the reversed phase separation of high molecular weight compounds such as monoclonal antibodies with molecular weight of 150 kDa.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service