Skip to Content
Merck
All Photos(1)

Key Documents

59578-U

Supelco

Discovery® RP-Amide C16 (5 µm) HPLC Columns

L × I.D. 2 cm × 3 mm Supelguard Guard Cartridge, pkg of 2 ea, Guard Cartridge holder required for use

Sign Into View Organizational & Contract Pricing

Select a Size

2 EA
$461.00

$461.00


Check Cart for Availability


Select a Size

Change View
2 EA
$461.00

About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

$461.00


Check Cart for Availability

Product Name

Discovery® RP-AmideC16 Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 3 mm

material

stainless steel column

Quality Level

Agency

suitable for USP L60

description

Supelguard Cartridge

product line

Discovery®

feature

endcapped

packaging

pkg of 2 ea

technique(s)

HPLC: suitable

L × I.D.

2 cm × 3 mm

surface area

200 m2/g

matrix

fully porous particle

matrix active group

amide, alkyl phase

particle size

5 μm

pore size

180 Å

operating pH

2-8

application(s)

food and beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Annamaria Jakab et al.
Journal of pharmaceutical and biomedical analysis, 27(6), 913-921 (2002-02-12)
Retention parameters of 45 different barbituric acid derivatives were determined on an amide embedded RP silica column (Discovery RP-AmideC16) using non-buffered water-acetonitrile eluent systems. Linear correlation were calculated between the logarithm of the capacity factor and the acetonitrile concentration in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service