Skip to Content
Merck
All Photos(1)

Key Documents

58702

PTFE Tubing

L × O.D. × I.D. 10 ft × 1/16 in. × 0.012 in.

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$85.60

$85.60


Available to ship on16 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 EA
$85.60

About This Item

UNSPSC Code:
41115700

$85.60


Available to ship on16 April 2025Details


Request a Bulk Order

packaging

pkg of 1 ea

parameter

500 psi max. pressure

L × O.D. × I.D.

10 ft × 1/16 in. × 0.012 in.

Looking for similar products? Visit Product Comparison Guide

General description

Economical, flexible PTFE tubing is ideal for use at pressures up to 500 psi (35 kg/cm2). Use in automatic analyzer, postcolumn reaction, preparative scale systems, in-stream sampling devices, and when monitoring physiologically important compounds.

The maximum recommended operating temperature for PTFE is 200 °C, but short term exposure to higher temperatures seldom causes damage.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service