581384-U
BIOshell lgG 1000 Å C4, 2.7 μm HPLC Column
column L × I.D. 15 cm × 1.5 mm
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
stainless steel hardware
Quality Level
Agency
suitable for USP L26
description
Shell thickness (0.5 μm)
Solid core (1.7 μm)
feature
endcapped
extent of labeling
0.6% carbon loading
parameter
90 °C max. temp.
column L × I.D.
15 cm × 1.5 mm
surface area
22 m2/g
matrix
spherical silica particle platform
matrix active group
C4 (butyl) phase
pore size
1000 Å
operating pH range
2-9
Looking for similar products? Visit Product Comparison Guide
General description
BIOshell IgG 1000Å C4 is a high-speed, high-performance liquid chromatography column based on a new wide-pore (1000Å) Fused-Core® particle design. The Fused-Core particle provides a thin porous shell of high-purity silica surrounding a solid silica core. This particle design exhibits very high column efficiency due to the shallow diffusion paths in the 0.5-micron thin porous shell and the small overall particle size of 2.7-microns. The densely bonded, extensively endcapped dimethylbutyl stationary phase of BIOshell IgG 1000Å C4 provides a stable, reversed-phase packing that can be used for separating high molecular weight compounds such as proteins.
Application
Legal Information
Fused-Core is a registered trademark of Advanced Materials Technology, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service