Skip to Content
Merck
All Photos(1)

Key Documents

58061

Supelco® Mobile Phase Filtration Apparatus 1

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$692.00

$692.00


Available to ship on17 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 EA
$692.00

About This Item

UNSPSC Code:
41115700

$692.00


Available to ship on17 April 2025Details


Request a Bulk Order

description

starter kit

Quality Level

packaging

pkg of 1

General description

Connects to 1000 mL sidearm flask

Includes:
  • 250 mL glass reservoir
  • Funnel base and stopper
  • Clamp
  • Stainless steel holder and screen
  • 10 PTFE gaskets
  • 50 Nylon 66 filters (47 mm, 0.45 μm pores).
·
Flask not included.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Supelco is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patrícia A B Ramos et al.
Antioxidants (Basel, Switzerland), 8(12) (2019-12-06)
Salix spp. have been exploited for energy generation, along with folk medicine use of bark extracts for antipyretic and analgesic benefits. Bark phenolic components, rather than salicin, have demonstrated interesting bioactivities, which may ensure the sustainable bioprospection of Salix bark.

Articles

Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service