Skip to Content
Merck
All Photos(1)

Key Documents

57494-U

Supelco

Supel Swift HLB 96-Well solid phase extraction (SPE) Plate

bed weight 30 mg, pk of 1 ea

Synonym(s):

Supel Swift HLB SPE 96-Well Plate

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$540.00

$540.00


Available to ship on07 April 2025Details


Request a Bulk OrderRequest a Sample

Select a Size

Change View
1 PKG
$540.00

About This Item

UNSPSC Code:
41104018
NACRES:
NB.21

$540.00


Available to ship on07 April 2025Details


Request a Bulk OrderRequest a Sample

Product Name

Supel Swift HLB SPE 96-Well Plate, bed weight 30 mg, pk of 1 ea

description

detection: MS friendly

Quality Level

composition

bed weight, 30 mg

packaging

pk of 1 ea

technique(s)

solid phase extraction (SPE): suitable

surface area

425-500 m2/g

matrix

polymeric HLB base material

particle size

40-75 μm (>90%)

pore size

80-210 Å pore size

Looking for similar products? Visit Product Comparison Guide

General description

Supel Swift HLB is a co-polymer solid phase extraction (SPE) phase composed of both hydrophilic and lipophilic functional groups. This phase is ideal for extracting a broad range of compounds from aqueous samples, including food and environmental samples, as well as biological samples (eg. serum and plasma). Because of the hydrophilic and lipophilic balanced (HLB) property, the polymer material can selectively retain a wide range of compounds with different polarities and LogP values. Traditional HLB SPE uses a five-step process for sample preparation that normally includes the conditioning of sorbents with an organic solvent followed by an equilibration step to return sorbents to sample loading conditions. The product design allows for direct sample loading onto the sorbent bed, thereby eliminating the conditioning step altogether. This product feature can both save time and simplify sample preparation, which may also serve to reduce sample processing errors.
The 96-well format is perfect for medium to high-throughput laboratories performing large numbers of samples at a time and is also amenable to automation.

Product Data Sheet

Application


  • Simultaneous determination of twelve neonicotinoids and six metabolites in human urine with isotope-dilution UPLC-Q-Orbitrap HRMS: This study utilizes the Supel Swift HLB SPE 96-Well Plate to develop a method for detecting neonicotinoids and their metabolites in human urine. The high-throughput solid-phase extraction (SPE) combined with ultra-performance liquid chromatography coupled with quadrupole Orbitrap high-resolution mass spectrometry (UPLC-Q-Orbitrap HRMS) showcases the plate′s efficiency in bioanalytical applications (Zhang et al., 2024).

  • Determination of sixteen antibiotics and four β-agonists in human urine samples using ultra-performance liquid chromatography-tandem mass spectrometry based on high-throughput automatic solid-phase extraction: This research employs the Supel Swift HLB SPE 96-Well Plate to extract multiple antibiotics and β-agonists from urine samples. The integration of automatic SPE with UPLC-MS/MS enhances the sensitivity and specificity of the method, demonstrating the plate′s application in pharmaceutical and clinical studies (Li et al., 2023).

  • Determination of phenoxyacetic herbicides, metabolites of organophosphorus and pyrethroid pesticides in human urine using solid phase extraction coupled with ultra-performance liquid chromatography-tandem mass spectrometry: This paper highlights the use of the Supel Swift HLB SPE 96-Well Plate for extracting various pesticides and their metabolites from urine samples. The SPE method′s high throughput and efficiency make it ideal for environmental and toxicological studies (Zhang et al., 2023).

  • Application of 96-well plate SPE method for analysis of persistent organic pollutants in low volume blood serum samples: The study applies the Supel Swift HLB SPE 96-Well Plate for analyzing persistent organic pollutants in blood serum. The method′s high sensitivity and low sample volume requirements make it valuable for biomonitoring and environmental health research (Palát et al., 2022).

Legal Information

Supel is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Supel™ Swift HLB SPE enhances solid phase extraction efficiency with faster flow rates and minimal signal interference.

Validate urine sample cleanup with HLB SPE for opioid analysis via LC-MS/MS using the Supel Swift HLB 96-well plate.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service