Skip to Content
Merck
All Photos(1)

Key Documents

57491-U

Supelco

Supel Swift HLB SPE Tubes

weight 200 mg (bed), volume 6 mL, pk of 30 ea

Synonym(s):

polymeric general SPE cartidge tube

Sign Into View Organizational & Contract Pricing

Select a Size

30 PKG
$243.00

$243.00


Available to ship on22 April 2025Details


Request a Bulk OrderRequest a Sample

Select a Size

Change View
30 PKG
$243.00

About This Item

UNSPSC Code:
41104018
NACRES:
NB.21

$243.00


Available to ship on22 April 2025Details


Request a Bulk OrderRequest a Sample

Agency

suitable for EPA 537.1
suitable for ISO 25101
suitable for ISO/CEN 15968-2010

Quality Level

description

detection: MS friendly

product line

Supel

packaging

pk of 30 ea

technique(s)

solid phase extraction (SPE): suitable

surface area

425-500 m2/g

volume

6 mL

weight

200 mg (bed)

particle size

40-75 μm (>90%)

pore size

80-210 Å pore size

application(s)

PFAS testing

Looking for similar products? Visit Product Comparison Guide

General description

Supel Swift HLB is a co-polymer solid phase extraction (SPE) phase composed of both hydrophilic and lipophilic functional groups. This phase is ideal for extracting a broad range of compounds from aqueous samples, including food and environmental samples, as well as biological samples (eg. serum and plasma). Because of the hydrophilic and lipophilic balanced (HLB) property, the polymer material can selectively retain a wide range of compounds with different polarities and LogP values. Traditional HLB SPE uses a five-step process for sample preparation that normally includes the conditioning of sorbents with an organic solvent followed by an equilibration step to return sorbents to sample loading conditions. The product design allows for direct sample loading onto the sorbent bed, thereby eliminating conditioning of the cartridge. This product feature can both save time and simplify sample preparation, which may also serve to reduce sample processing errors.

Legal Information

Supel is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Supel™ Swift HLB SPE enhances solid phase extraction efficiency with faster flow rates and minimal signal interference.

LC-MS/MS-based determination of 19 sulfonamides, 13 quinolones, and 3 tetracycline drug residues in pork meat samples using Supel™ Swift HLB for extraction.

Related Content

FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.

Questions

  1. As per the protocol only 200 ul sample is to be loaded. What if i load 1ml sample

    1 answer
    1. This product is optimized to perform best with the recommended 200 µl sample volume. A 1 mL sample would overload the tube, likely impacting the quality of results.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service