Skip to Content
Merck
All Photos(1)

Key Documents

57035-U

Supelco

SPE Manifold Gauge/Bleed Valve

for use with Visiprep 12-port and 24-port models

Synonym(s):

V.M.VACUUM GAUGE AND VALVE

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$550.00

$550.00


Available to ship on23 April 2025Details


Request a Bulk OrderRequest a Sample

Select a Size

Change View
1 EA
$550.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.21

$550.00


Available to ship on23 April 2025Details


Request a Bulk OrderRequest a Sample

technique(s)

solid phase extraction (SPE): suitable

Quality Level

application(s)

food and beverages

compatibility

for use with Visiprep 12-port and 24-port models

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service