Skip to Content
Merck
All Photos(1)

Key Documents

55417-U

Supelco

Supel QuE

Z-Sep with MgSO4 Tube, centrifuge tube volume 2 mL, pack of 100 ea

Synonym(s):

QuEChERS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115712

composition

Magnesium Sulfate, 150 mg
Z-Sep, 50 mg

packaging

pack of 100 ea

technique(s)

QuEChERS: suitable

centrifuge tube volume

2 mL

application(s)

food and beverages

Looking for similar products? Visit Product Comparison Guide

General description

Dispersive SPE (dSPE), often referred to as the "QuEChERS" method (Quick, Easy, Cheap, Effective, Rugged, and Safe), is modern sample prep technique that is becoming increasingly popular in the area of multi-residue pesticide analysis in food and agricultural products.

Using the QuEChERS method, food/agricultural samples are first extracted with an aqueous miscible solvent (e.g., acetonitrile) in the presence of high amounts of salts (e.g., sodium chloride and magnesium sulfate) and/or buffering agents (e.g. citrate) to induce liquid phase separation and stabilize acid and base labile pesticides, respectively. Upon shaking and centrifugation, an aliquot of the organic phase is subjected to further cleanup using SPE. Unlike traditional methods using SPE tubes, in dispersive SPE, cleanup is facilitated by mixing bulk amounts of SPE (e.g., Supelclean PSA, ENVI-Carb, and/or Discovery DSC-18) with the extract. After sample cleanup, the mixture is centrifuged and the resulting supernatant can either be analyzed directly or can be subjected to minor further treatment before analysis.

Supelco carries a line of vials and centrifuge tubes containing pre-determined amounts of salts and SPE sorbents to support the most common method configurations used today.

Other Notes

Supel QuE Z-Sep products enhance sample cleanup for complex matrices by effectively removing more fat and color from sample extracts than traditional phases for QuEChERS methods. By eliminating problematic matrix interferences, Z-Sep products provide more robust LC-MS and GC-MS methods. This proprietary technology can replace C18 and PSA in your current methods without additional method development.

Z-Sep+ is recommended for cleanup of samples containing greater than 15% fat.
Z-Sep/C18 is recommended for cleanup of samples containing less than 15% fat.
Z-Sep is recommended for cleanup of samples prior to analysis of hydrophobic analytes.

Legal Information

Certain uses of this product may be covered by the claims of one or more pending patent applications. For licensing inquiries please contact [email protected]
AOAC is a registered trademark of AOAC International
Supel is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Łukasz Rajski et al.
Journal of chromatography. A, 1304, 109-120 (2013-07-23)
Several extraction methods were evaluated in terms of recoveries and extraction precision for 113 pesticides in avocado: QuEChERS with various d-SPE clean-ups (Z-Sep, Z-Sep+, PSA+C18 and silica), miniLuke and ethyl acetate. Extracts were analysed using liquid chromatography coupled with triple
Pol Herrero et al.
Journal of chromatography. A, 1339, 34-41 (2014-03-25)
A Quick, Easy, Cheap, Effective, Rugged and Safe (QuEChERS) extraction method followed by liquid chromatography-(Orbitrap) high resolution mass spectrometry was developed for the simultaneous determination of five benzotriazole, four benzothiazole and five benzenesulfonamide derivates in sewage sludge. While the method
Lianzhu Wang et al.
Se pu = Chinese journal of chromatography, 33(5), 501-507 (2015-09-22)
A fast method based on QuEChERS methodology for the simultaneous determination of 32 sulfonylurea herbicide residues in sweet corns and green soybeans was developed using liquid chromatography-tandem mass spectrometry (LC-MS/MS). The clean-up effects of three dispersive sorbents were evaluated in
Tomasz Tuzimski et al.
Journal of AOAC International, 100(6), 1671-1680 (2017-07-15)
In this study, a simple, cost-effective, and sensitive HPLC diode-array detection method was developed for the simultaneous determination of six different 5-nitroimidazoles [metronidazole, 2-hydroxymethyl-1-methyl-5-nitro-1H-imidazole, dimetridazole (DMZ), ronidazole, ornidazole, and ipronidazole] in bovine milk samples. A QuEChERS-based sample preparation procedure was
Hui Li et al.
Journal of chromatography. A, 1528, 53-60 (2017-11-07)
Herein, an accurate and reliable isotope-labelled internal standard method was developed and validated for simultaneous determination of three polar pesticide adjuvants, namely 2-pyrrolidone, N-methyl-2-pyrrolidone, and N-ethyl-2-pyrrolidone in plant-derived agro-products. Matrices, including apple, cabbage, tomato, cucumber, rice, and wheat were extracted

Articles

Assessment of clean-up sorbents for extracting PAHs from salmon and PCBs from milk, optimized for GC analysis with MS and ECD detection.

Related Content

Z-Sep sorbents improve fat and pigment extraction in QuEChERs solid phase extraction for sample preparation.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service