Key Documents
55076
Upchurch Tubing Adapter
Select a Size
1 EA
$27.50
$27.50
Check Cart for Availability
Request a Bulk Order
Select a Size
Change View
1 EA
$27.50
About This Item
UNSPSC Code:
41115700
$27.50
Check Cart for Availability
Request a Bulk Order
material
natural PEEK adapter (one-piece design)
packaging
pkg of 1 ea
fitting
female Luer (lock rim)
10-32 female
General description
Upchurch Tubing Adapter, made of PEEK, is natural in color. It is used to connect membrane adsorber units to a workstation.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Choose from one of the most recent versions:
Certificates of Analysis (COA)