Skip to Content
Merck
All Photos(1)

Key Documents

52052

Supelco

Trimethylolpropane trimethacrylate

analytical standard, contains 0.01-0.04% monomethyl ether hydroquinone as stabilizer

Synonym(s):

TMPTMA

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$145.00

$145.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$145.00

About This Item

Linear Formula:
[H2C=C(CH3)CO2CH2]3CC2H5
CAS Number:
Molecular Weight:
338.40
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$145.00


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

vapor density

>1 (vs air)

vapor pressure

<0.01 mmHg ( 20 °C)

Assay

≥95.0% (GC)

shelf life

limited shelf life, expiry date on the label

contains

0.01-0.04% monomethyl ether hydroquinone as stabilizer

refractive index

n20/D 1.470-1.474
n20/D 1.472 (lit.)

density

1.06 g/mL at 25 °C (lit.)

SMILES string

CCC(COC(=O)C(C)=C)(COC(=O)C(C)=C)COC(=O)C(C)=C

InChI

1S/C18H26O6/c1-8-18(9-22-15(19)12(2)3,10-23-16(20)13(4)5)11-24-17(21)14(6)7/h2,4,6,8-11H2,1,3,5,7H3

InChI key

OKKRPWIIYQTPQF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service