Skip to Content
Merck
All Photos(1)

Key Documents

50982

Triethyl phosphite

analytical standard

Synonym(s):

P(EtO)3, P(OEt)3

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$126.00

$126.00


Estimated to ship on13 April 2025


Request a Bulk Order

Select a Size

Change View
1 ML
$126.00

About This Item

Linear Formula:
(C2H5O)3P
CAS Number:
Molecular Weight:
166.16
Beilstein:
956578
MDL number:
UNSPSC Code:
12352108
NACRES:
NA.21

$126.00


Estimated to ship on13 April 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥97.5% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.412-1.414
n20/D 1.413 (lit.)

bp

156 °C (lit.)

density

0.969 g/mL at 25 °C (lit.)

SMILES string

CCOP(OCC)OCC

InChI

1S/C6H15O3P/c1-4-7-10(8-5-2)9-6-3/h4-6H2,1-3H3

InChI key

BDZBKCUKTQZUTL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service