Skip to Content
Merck
All Photos(1)

Key Documents

8.00549

Sigma-Aldrich

Triethylphosphite

for synthesis

Synonym(s):

Triethylphosphite

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$52.80
500 ML
$112.00

$52.80


Only 1 left in stockDetails

Request a Bulk Order

Select a Size

Change View
100 ML
$52.80
500 ML
$112.00

About This Item

Empirical Formula (Hill Notation):
C6H15O3P
CAS Number:
Molecular Weight:
166.16
MDL number:
UNSPSC Code:
12352108
EC Index Number:
204-552-5
NACRES:
NA.22

$52.80


Only 1 left in stockDetails

Request a Bulk Order

vapor pressure

<6 hPa ( 20 °C)

Quality Level

Assay

≥97.0% (GC)

form

liquid

autoignition temp.

250 °C

potency

2800 mg/kg LD50, skin (Rabbit)

expl. lim.

3.75-42.5 % (v/v)

bp

156-158 °C/1013 hPa

mp

-112 °C

transition temp

flash point 52 °C

density

0.956 g/cm3 at 20 °C

storage temp.

no temp limit

SMILES string

P(OCC)(OCC)OCC

InChI

1S/C6H15O3P/c1-4-7-10(8-5-2)9-6-3/h4-6H2,1-3H3

InChI key

BDZBKCUKTQZUTL-UHFFFAOYSA-N

Application


  • Mechanism of acylative oxidation-reduction-condensation reactions: Research details the use of triethylphosphite as a stoichiometric reductant in acylative oxidation-reduction-condensation reactions, utilizing benzoisothiazolones as oxidants. This study provides insights into the reaction mechanisms and potential applications in synthetic chemistry (Gangireddy et al., 2017).

  • Synthesis and antitumor activity of oxazaphosphinane derivatives: A study on the efficient synthesis of novel oxazaphosphinane derivatives using triethylphosphite, exploring their antitumor activities. The research includes X-ray crystallography, DFT studies, and molecular docking to evaluate the potential medical applications (Bahadi et al., 2023).

  • Reductive Ireland-Claisen Rearrangements: Triethylphosphite is highlighted in copper-catalyzed reductive Ireland-Claisen rearrangements of propargylic acrylates and allylic allenoates, demonstrating its role in facilitating complex molecular transformations in organic synthesis (Guo et al., 2022).

  • Desulfurization under UV light: Triethylphosphite is used in the synthesis of deoxyglycosides through desulfurization processes under UV light, showcasing its utility in glycoscience and the development of new glycoconjugates for various applications (Ge et al., 2017).

  • Detection and identification of volatile trialkylphosphites: A derivatization strategy employing triethylphosphite for the detection and identification of volatile trialkylphosphites using liquid chromatography-online solid phase extraction and offline nuclear magnetic resonance spectroscopy, highlighting its analytical applications (Mazumder et al., 2015).

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.955 - 0.957
Identity (IR): passes test

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Flam. Liq. 3 - Skin Sens. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

129.2 °F - closed cup

Flash Point(C)

54 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service