Key Documents
509736
SGE Syringes for PerkinElmer® Autosystem
volume 5 μL, 5F-PE-0.47, needle size 26 ga, needle L 70 mm
Select a Size
1 EA
$182.00
$182.00
Check Cart for Availability
Request a Bulk Order
Select a Size
Change View
1 EA
$182.00
About This Item
UNSPSC Code:
41122001
$182.00
Check Cart for Availability
Request a Bulk Order
material
cone tip stainless steel needle (point style 1)
needle size
26 ga
description
color (lime)
liquid syringe, fixed needle
feature
needle type LTN (fixed)
packaging
pkg of 1 ea
manufacturer/tradename
SGE 001953
needle L
70 mm
volume
5 μL
Legal Information
PerkinElmer is a registered trademark of PerkinElmer Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)