Skip to Content
Merck
All Photos(1)

Key Documents

504262

Supelco

Supelguard HPLC Guard Cartridge Holder

Direct-Connect (Swivel-type), for use with Supelguard cartridges (2 cm L. x 2.1 mm I.D.)

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$238.00

$238.00


Check Cart for Availability


Select a Size

Change View
1 EA
$238.00

About This Item

UNSPSC Code:
41115700

$238.00


Check Cart for Availability

Product Name

Supelguard Guard Cartridge Holder, Direct-Connect (Swivel-type), for use with Supelguard cartridges (2 cm L. x 2.1 mm I.D.)

material

stainless steel housing

fitting

10-32 female UNF (Valco compatible)
5/16-40 female UNF (threads directly onto HPLC column)

compatibility

for use with Supelguard cartridges (2 cm L. x 2.1 mm I.D.)

Looking for similar products? Visit Product Comparison Guide

Footnote

For 2.1 mm I.D. Supelco columns.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service