Key Documents
502685
Upchurch Precolumn MicroFilter
for use with 1/16 in. O.D. tubing, stainless steel frit
Select a Size
1 EA
$237.00
$237.00
Only 0 left in stock
Request a Bulk Order
Select a Size
Change View
1 EA
$237.00
About This Item
UNSPSC Code:
41115700
$237.00
Only 0 left in stock
Request a Bulk Order
material
stainless steel frit
packaging
pkg of 1 ea
fitting
10-32 female Upchurch inlet fitting for 1/16 in. O.D. tubing
10-32 male Upchurch outlet fitting for 1/16 in.
compatibility
for use with 1/16 in. O.D. tubing
General description
PEEK housing with in-line filter connects capillary tubing (using Microtight tubing sleeves) or 1/16 in. O.D. tubing to female 10-32 fitting. Swept volume of 0.3 μL. Includes 5 × 0.5 μm frits (one installed).
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Choose from one of the most recent versions:
Certificates of Analysis (COA)