Skip to Content
Merck
All Photos(1)

Key Documents

502677

Upchurch Precolumn MicroFilter

for use with 0.025 in. O.D. tubing, PEEK frit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

material

PEEK frit

packaging

pkg of 1 ea

fitting

10-32 female Upchurch inlet fitting for 0.025 in. (using Microtight tubing sleeves)
10-32 male Upchurch outlet fitting for 1/16 in. O.D. tubing

compatibility

for use with 0.025 in. O.D. tubing

Looking for similar products? Visit Product Comparison Guide

General description

PEEK housing with in-line filter connects capillary tubing (using Microtight tubing sleeves) or 1/16 in. O.D. tubing to female 10-32 fitting. Swept volume of 0.3 μL. Includes 5 × 0.5 μm frits (one installed).

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service