Key Documents
500410
Supelco
Stand for SCOTTY® 48 Cylinder
Select a Size
1 EA
$177.00
$177.00
Check Cart for Availability
Request a Bulk Order
Select a Size
Change View
1 EA
$177.00
About This Item
UNSPSC Code:
12352200
$177.00
Check Cart for Availability
Request a Bulk Order
packaging
pkg of 1 ea
application(s)
environmental
General description
Stabilizes your cylinder on a benchtop or other flat surface. This item will hold cylinders with a four-inch diameter.
Application
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Legal Information
SCOTTY is a registered trademark of Air Liquide
Choose from one of the most recent versions:
Certificates of Analysis (COA)