Skip to Content
Merck
All Photos(1)

Key Documents

48527

Supelco

trans-1,2-Dichloroethene

analytical standard

Synonym(s):

trans-1,2-Dichloroethylene, trans-1,2-Dichloroethene, trans-Acetylene dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH=CHCl
CAS Number:
Molecular Weight:
96.94
Beilstein:
1420761
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

vapor pressure

5.16 psi ( 20 °C)

expl. lim.

12.8 %

packaging

ampule of 5000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.446 (lit.)

bp

48 °C (lit.)

mp

−50 °C (lit.)

density

1.257 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

Cl\C=C\Cl

InChI

1S/C2H2Cl2/c3-1-2-4/h1-2H/b2-1+

InChI key

KFUSEUYYWQURPO-OWOJBTEDSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

42.8 °F - closed cup

Flash Point(C)

6.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Timothy E Mattes et al.
FEMS microbiology reviews, 34(4), 445-475 (2010-02-12)
Extensive use and inadequate disposal of chloroethenes have led to prevalent groundwater contamination worldwide. The occurrence of the lesser chlorinated ethenes [i.e. vinyl chloride (VC) and cis-1,2-dichloroethene (cDCE)] in groundwater is primarily a consequence of incomplete anaerobic reductive dechlorination of
Florian Schevenels et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 19(13), 4335-4343 (2013-01-22)
Highly functionalised benzofurans have been prepared from ortho-hydroxyphenones and 1,1-dichloroethylene. The key intermediate, a chloromethylene furan, smoothly rearranged into the corresponding benzofuran carbaldehyde under acidic conditions. Some mechanistic investigations have been performed and several biologically active benzofurans have been synthesised.
Darlene D Wagner et al.
BMC genomics, 13, 200-200 (2012-05-24)
Geobacter lovleyi is a unique member of the Geobacteraceae because strains of this species share the ability to couple tetrachloroethene (PCE) reductive dechlorination to cis-1,2-dichloroethene (cis-DCE) with energy conservation and growth (i.e., organohalide respiration). Strain SZ also reduces U(VI) to
Federico Aulenta et al.
Bioresource technology, 101(24), 9728-9733 (2010-08-17)
Quinone moieties in humic substances have previously been shown to serve as extracellular electron acceptors in different metabolic pathways. Here we show that the humic acid analogue antraquinone-2,6-disulfonate (AQDS) can also serve as an electron donor in the microbial reductive
Ramona Darlington et al.
Chemosphere, 90(8), 2226-2232 (2012-10-30)
A fractured sandstone aquifer at an industrial site is contaminated with trichloroethene to depths greater than 244 m. Field data indicate that trichloroethene is undergoing reduction to cis-1,2-dichloroethene (cDCE); vinyl chloride and ethene are present at much lower concentrations. Transformation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service